Learn More
Hettich Lab Technology™ ROTINA 380 Benchtop Centrifuge
Product Code. 10075563
Change view
Click to view available options
Capacity:
4 x 290 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
10075563 4 x 290 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10075563 Supplier Hettich Lab Technology™ Supplier No. 1701
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
ROTINA 380 Benchtop Centrifuge is a high-performance, high-speed, space-saving unit supporting a wide array of accessories for multiple applications in just one rotor.
  • Ideal for medium to HTP labs processing large quantities of blood collection tubes in swing-out and angle rotors
  • A multitude of containers for cell culture can be spun as well as 50 mL conical tubes for the passaging of cells
  • A vast variety of plates and racks for solid phase assays or for cell culture assays can be processed
  • Excellent for use in facilities such as hospitals, hematological labs, small blood centers, veterinary labs and more
Specifications
Capacity 4 x 290 mL
Max. RCF 24,400 x g
Max. Speed 15,000 rpm
Refrigerated Yes
Model 380
Run Time 1 sec. to 99 hr., 59 min., 59 sec., Infinite Continuous Run, Short Cycle Mode
Warranty 5 Years
Dimensions (HWD) 418 x 457 x 600 mm
Net Weight 51 kg
Electrical Requirements 220/240 V
Product Type Table Top Centrifuge