Learn More
Sartorius Table Paper Boards
Shop All Sartorius Products
Click to view available options
Dimensions (L x W):
210 x 297 mm (8.3 x 11.7 in.)
580 x 580 mm (22.8 x 22.8 in.)
Unit Size:
Pack of 100
Pack of 200
Description
Filter Boards
| Grade | Basis weight g/m2 | Thickness in mm |
| 0.90 | 420 | 1.30 |
| 1220 | 475 | 1.10 |
| 1339 | 315 | 0.63 |
| 151 | 460 | 0.90 |
| 152 | 380 | 0.90 |
| 152 A | 340 | 1.00 |
| 1522 | 280 | 0.60 |
| 157 | 700 | 1.80 |
| 1600 | 250 | 0.50 |
| 1632 | 150 | 0.63 |
| 167 360 | 360 | 0.90 |
| 511 | 490 | 1.25 |
| 64 | 100 | 0.22 |
| A 140 | 140 | 0.25 |
| A 250 | 250 | 0.40 |
| C 160 | 160 | 0.30 |
| C 250 | 250 | 0.43 |
| C 251 | 250 | 0.55 |
| C 300 | 300 | 0.52 |
| C 350 | 350 | 0.63 |
| C 350 L | 360 | 0.75 |
| C 450 | 450 | 0.95 |
| K 12 | 520 | 1.50 |
| LF 1 | 360 | 0.90 |
| M 270 | 270 | 0.62 |
| M 600 | 600 | 1.60 |
| S 165 | 165 | 0.33 |
| S 500 | 500 | 1.00 |
| SB 2 | 155 | 0.34 |
| SEK 550 | 550 | 1.25 |
| SEK 770 | 770 | 1.00 |
Specifications
Specifications
| Format | Sheet |
| Grade | H182 |
| Type | Filter Board |
| Dimensions (L x W) | 580 x 580 mm (22.8 x 22.8 in.) |
| Fit for Purpose | Certified Quality |