Learn More
TAB1 Mouse anti-Human, Unconjugated, Clone: 1E10D12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67020-1-IG-150UL
 View more versions of this product
Product Code. 16839395
  • 4836.10 NOK / 150µL
Description
The TAB1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TAB1. This antibody recognizes human, mouse, rat antigen. The TAB1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TAB1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
1E10D12
Unconjugated
150 μL
10454
IgG1