Learn More
TAB1 Mouse anti-Human, Unconjugated, Clone: 1E10D12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67020-1-IG-150UL
This item is not returnable.
View return policy
Description
The TAB1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TAB1. This antibody recognizes human, mouse, rat antigen. The TAB1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TAB1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, ELISA | |
| 1E10D12 | |
| Unconjugated | |
| 150 μL | |
| 10454 | |
| IgG1 |