Description
- Features a brown UV-protection coat
- Brown glass bottle for titrant
- GL 45 thread size
- Includes S 40 bottle adapter, hoses, drip tube, and titration tip
Specifications
Specifications
| Type | Exchange Unit |
| Includes | Reagent bottle |
| Volume (Metric) | 20 mL |