Learn More
T56-LIMKi, MedChemExpress
Product Code. 16483717
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16483717
Brand: Medchem Express HY19352
MedChemExpress T56-LIMKi is a selective inhibitor of LIMK2; inhibits the growth of Panc-1 cells with an IC50 of 35.2 μM.
T56-LIMKi is a selective inhibitor of LIMK2
Specifications
Chemical Name or Material T56-LIMKi
Physical Form Solid
CAS 924473-59-6
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H14F3N3O3
For Use With (Application) Cancer-Kinase/protease
Synonym T5601640
Solubility Information DMSO : 100 mg/mL (256.85 mM; Need ultrasonic)
SMILES O=C(C1=CC(C)=NO1)NC2=CC=CC(C(NC3=CC=CC(C(F)(F)F)=C3)=O)=C2
Molecular Weight (g/mol) 389.33
Formula Weight 389.33
Percent Purity 99.07%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.