Learn More
T0901317, MedChemExpress
Product Code. 16444078
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16444078
Brand: Medchem Express HY10626
MedChemExpress T0901317 is an orally active and highly selective LXR agonist with an EC50 of 20 nM for LXRα. T0901317 activates FXR with an EC50 of 5 μM. T0901317 is RORα and RORγ dual inverse agonist with Ki values of 132 nM and 51 nM, respectively. T0901317 induces apoptosis and inhibits the development of atherosclerosis in low-density lipoprotein (LDL) receptor-deficient mice.
T0901317 is an orally active and highly selective LXR agonist with an EC50 of 20 nM for LXRα. T0901317 activates FXR with an EC50 of 5 μM. T0901317 is RORα and RORγ dual inverse agonist with Ki values of 132 nM and 51 nM, respectively. T0901317 induces apoptosis and inhibits the development of atherosclerosis in low-density lipoprotein (LDL) receptor-deficient mice.
Specifications
Chemical Name or Material T0901317
CAS 293754-55-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H12F9NO3S
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (207.76 mM; Need ultrasonic)
SMILES OC(C(F)(F)F)(C(F)(F)F)C1=CC=C(N(CC(F)(F)F)S(C2=CC=CC=C2)(=O)=O)C=C1
Molecular Weight (g/mol) 481.33
Formula Weight 481.33
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.