Learn More
Grant Instruments™ T-Clamp
Description
The Grant T-Clamp allows a Grant heating circulator to be attached to virtually any vertical sided vessel.
Grant heated circulators are suitable for use with Grant stainless steel and plastic tanks. With the addition of a clamp (T-Clamp) they can also be attached to virtually any vertical sided tank with a maximum wall thickness of 35mm for rectangular tanks, 30mm for circular tanks (300mm diameter), and a capacity of up to 50 litres. Minimum and maximum temperatures achievable are dependent upon the tank insulation and minimum operating temperature depends on the accessory cooling device.
Specifications
Specifications
| Capacity (Metric) | 50 L |
| For Use With (Application) | Allows heating circulator to be attached to virtually any vertical sided vessel with a maximum wall |