Learn More
Sampling Systems™ T Bar Handle for Unit Dose Sleeve Sampler
Shop All Sampling Systems Products
Click to view available options
Length (Metric):
600 mm
Unit Size:
Each
Description
- For sampling small volume powders and granules
- Chemically resistant 316L stainless steel construction
- Different sample sizes can be taken using the same sampler
- Allows for easy use of sampler
Specifications
Specifications
| Material | Stainless Steel (316L) |
| Description | T Bar Handle |
| Handle Type | T-shape |
| For Use With (Equipment) | Unit Dose Sleeve Sampler |
| Length (Metric) | 600 mm |