Learn More
Velp Scientifica™ System Support
Product Code. p-8012699 Shop All Velp Products
Click to view available options
For Use With (Equipment):
DK20 and DK42/26
Unit Size:
Each
Product Code. 13445509
Brand: Velp Scientifica™ A00000190
Only null left
Add to basket
Add to basket
Velp Scientifica™ System Support is designed for use with a variety of Kjeldahl Digestion Units.
  • Required accessory with DK Series Digestion Unit
Specifications
Product Type System Support
For Use With (Equipment) DK20 and DK42/26