Learn More
CELLPATH SYSTEM III BIOPSY CASSETTE
Click to view available options
Material:
Loose
Threaded
Unit Size:
Pack of 2000
Pack of 500
Description
- The locking mechanism of the cassette ensures maximum tissue security during processing.
- The cassette has a precisely engineered writing surface which optimises the print quality on inkjet and thermal printers, including the Primera, Leica and Sakura Printers.
- Key Benefits:
- Compatible with Primera, Leica and Sakura Printers
- Optimised writing surface
- Increased reagent flow
- Integral fixed frangible lid
- High rigidity and stability
Specifications
Specifications
| Color | White |
| Material | Loose |
| Product Type | Cassette |
Product Content Correction
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur