Learn More
CELLPATH SYSTEM II+ 4 WELL CASSETTE
Product Code. p-200113608
Click to view available options
Material:
Hoppered
Loose
Threaded
Unit Size:
Pack of 1000
Pack of 1500
Pack of 2000
Product Code. 30226259
Brand: CELLPATH EAM180203A
The System II+ 4 Well Cassette incorporates multiple wells into the cassette allows multiple biopsies to be processed simultaneously, significantly reducing user workload. They come with a separate white lid that is easily attached via the use of a hinge.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Specifications
Color White
Material Loose
Product Type Cassette