Learn More
Thermo Scientific™ Syringes for Thermo Scientific™ HPLC Instruments
Thermo Scientific™ Syringes for Thermo Scientific HPLC Instruments offer easy, accurate and reproducible injections.
Brand:  Thermo Scientific™ 365LLT81
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10197744
  • 2106.68 NOK / Each
Estimated Shipment: 12-06-2024
Description
Thermo Scientific™ Syringes for Thermo Scientific HPLC Instruments offer easy, accurate and reproducible injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
Specifications
Specifications
HPLC Autosampler Syringe
No
2.5 mL
AS3000, AS3500 Autosampler