Learn More
Hamilton™ 1000 Series Luer Lock Syringes
Click to view available options
Volume (Metric):
50 mL
Unit Size:
Each
Description
- 50mL volume
- For Metal or Kel-F Hubs
- Without flange
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Product Type | Syringe Needle |
| Max. Pressure | 6.9 bar |
| Max. Temperature (Metric) | 50°C |
| Model | 1050 |
| Quantity | 1/Pk. |
| Volume (Metric) | 50 mL |
| Connection Type | Luer Lock |