Learn More
Hamilton™ Custom Cemented Needle Syringes
Product Code. 10491245
Change view
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
25 mL
Product Type:
Gastight
Syringe Needle
Unit Size:
Each
Product Code. Volume (Metric) Product Type unitSize
10491245 25 μL Gastight Each
11518024 10 μL Gastight Each
10595705 100 μL Syringe Needle Each
10194402 100 μL Gastight Each
10281973 250 μL Syringe Needle Each
10201242 250 μL Gastight Each
10234272 25 mL Syringe Needle Each
10175122 50 μL Gastight Each
10349551 500 μL Syringe Needle Each
9 options
Product Code. 10491245
Brand: Hamilton™ 80208
The gas-tight version of the original 700 series syringes.
  • Ideal for dispensing volumes from 1 up to 500 μL
  • A variety of volumes, lengths, and gauges available
Specifications
Autoclavable Not Autoclavable
Needle Gauge Custom
Diameter (Metric) Inner 0.729 mm
Diameter (Metric) Outer 7.75 mm
Product Type Gastight
Max. Pressure 68.9 bar
Max. Temperature (Metric) 50°C
Model 1702
Min. Temperature (Metric) 10°C
Quantity 1 Ea.
Volume (Metric) 25 μL
Length (Metric) Needle Custom Needle
Needle Point Style Custom
Connection Type Cemented