Learn More
Syringe Filter with Hose Connector Adapter
For use with Twin hose connector
Brand: Pyrex 4541/04
807.50 NOK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.02820kg
Description
- Optional venting connector with a 0.2μm syringe filter
Specifications
Syringe Filter | |
Hose connector adapter |