Learn More
Socorex™ Dosys™ All-glass Syringes
Artikelnummer. 10131442
Ansicht ändern
Click to view available options
Volume (Metric):
0.1 to 1 mL
0.2 to 5 mL
0.5 to 2 mL
1 to 10 mL
1 to 20 mL
1 to 30 mL
1 to 50 mL
10 to 150 mL
10 to 200 mL
Connection Type:
Luer Lock
Luer Nozzle
Packungsgröße:
Each
Pack of 2
Pack of 3
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Volume (Metric) Connection Type Packungsgröße
10131442 1 to 50 mL Luer Lock Each
10477072 0.5 to 2 mL Luer Lock Pack of 3
10029761 1 to 20 mL Luer Lock Pack of 2
10039701 0.2 to 5 mL Luer Nozzle Pack of 3
10324932 1 to 20 mL Luer Nozzle Pack of 2
10161342 1 to 10 mL Luer Nozzle Pack of 3
10551713 0.2 to 5 mL Luer Lock Pack of 3
10385022 1 to 10 mL Luer Lock Pack of 3
11837253 0.1 to 1 mL Luer Nozzle Pack of 3
12663275 10 to 150 mL Luer Lock Each
12673275 10 to 200 mL Luer Lock Each
15562185 1 to 30 mL Luer Nozzle Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10131442 Lieferant Socorex™ Lieferanten-Nr. 155.0550
Estimated Shipment: 22-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Resistant borosilicate glass syringes can be sterilized and reused. Socorex™ Dosys™ All-glass Syringes provide a low-cost alternative to disposable plastic syringes and are easy to disassemble for cleaning.
  • Borosilicate glass construction.
  • Superior chemical, heat and shock resistance.
  • Low-cost alternative to disposable plastic syringes.
  • Precision-machined plunger and barrel.
  • Excellent fitting and liquid tightness (not gas tight).
  • Permanent, high visibility graduations.
  • Glass Luer and metal Luer lock nozzles available.
  • Can be sterilized up to 160°C / 320°F.
  • Very easy to disassemble and clean.
Spezifikation
Autoclavable Autoclavable
Material Glass, Glass (Barrel), Glass (Plunger), Metal (Tip)
Product Type Syringe
Graduations 2 mL
Quantity 1/ea.
Volume (Metric) 1 to 50 mL
Connection Type Luer Lock
Graduated Yes