Learn More
Socorex™ Dosys™ All-glass Syringes
Click to view available options
Volume (Metric):
0.1 to 1 mL
0.2 to 5 mL
0.5 to 2 mL
1 to 10 mL
1 to 20 mL
1 to 30 mL
1 to 50 mL
10 to 150 mL
10 to 200 mL
Connection Type:
Luer Lock
Luer Nozzle
Packungsgröße:
Each
Pack of 2
Pack of 3
Beschreibung
- Borosilicate glass construction.
- Superior chemical, heat and shock resistance.
- Low-cost alternative to disposable plastic syringes.
- Precision-machined plunger and barrel.
- Excellent fitting and liquid tightness (not gas tight).
- Permanent, high visibility graduations.
- Glass Luer and metal Luer lock nozzles available.
- Can be sterilized up to 160°C / 320°F.
- Very easy to disassemble and clean.
Spezifikation
Spezifikation
| Autoclavable | Autoclavable |
| Material | Glass, Glass (Barrel), Glass (Plunger), Metal (Tip) |
| Product Type | Syringe |
| Graduations | 1 mL |
| Quantity | 2/Pk. |
| Volume (Metric) | 1 to 20 mL |
| Connection Type | Luer Lock |
| Graduated | Yes |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur