Learn More
PURUS™ Synthetic Cleanroom Paper
Product Code. 15763579 Shop All Purus Products
Change view
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
Size:
A3
A4
Weight (Metric):
100 gsm
105 gsm
80 gsm
Quantity:
250 Sheets/Pk., 10 Packs/Cs.
250 Sheets/Pk., 5 Packs/Cs.
Unit Size:
Pack of 10
Pack of 250
Pack of 5
Product Code. Color Size Weight (Metric) Quantity unitSize
15763579 White A4 105 gsm 250 Sheets/Pk., 10 Packs/Cs. Pack of 250
15713579 Blue A4 80 gsm 250 Sheets/Pk., 10 Packs/Cs. Pack of 10
15743579 Green A4 80 gsm 250 Sheets/Pk., 10 Packs/Cs. Pack of 10
15753579 Pink A4 80 gsm 250 Sheets/Pk., 10 Packs/Cs. Pack of 10
15773579 White A4 80 gsm 250 Sheets/Pk., 10 Packs/Cs. Pack of 10
15783579 White A4 100 gsm 250 Sheets/Pk., 10 Packs/Cs. Pack of 10
15793579 Yellow A4 80 gsm 250 Sheets/Pk., 10 Packs/Cs. Pack of 10
15723579 Blue A3 80 gsm 250 Sheets/Pk., 5 Packs/Cs. Pack of 5
8 options
Product Code. 15763579
Brand: PURUS™ LCIW 2030C
Only null left
Add to basket
Add to basket
Cleanroom paper that excels in Tensile Strength. For writing, printing and photocopying in cleanroom environments, PURUS™ Cleanroom Paper provides excellent printability and heat toner adhesion and is recyclable.
  • Features high tensile strength and excellent heat resistance for enhanced copier and printing efficiency
  • Propriatary binding technology results in very low levels of particles, nonvolatile residue (NVR) and ion extractables
  • Unique precision cutting technology guarantees consistent sizing and cleanliness
  • Produced in ISO 4 (Class 10) cleanroom
  • Recyclable
  • Suitable for use in ISO 4 to 5 (Class 10 to Class 1000) cleanrooms
Specifications
Material Synthetic
Ruled No
Color White
Dimensions (L x W) 210 x 297 mm
Autoclavable Autoclavable
For Use With (Application) For Class 10 to Class 1000 cleanrooms, writing, printing and photocopying in clean environments
Plain paper Yes
Product Type Cleanroom Paper
Quantity 250 Sheets/Pk., 10 Packs/Cs.
Size A4
Weight (Metric) 105 gsm