Learn More
Corning™ Synthemax™ II Dissolvable Microcarriers
Shop All Corning Products
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
Description
- Sterile
- Ready-to-use
- Available in closed systems packaging to be used directly with bioreactors
- Additionally, the microcarriers are surface-treated to enhance cell attachment, maximizing cell yield and viability
- Consistency – USP Class VI polystyrene material provides a consistent platform
- Performance – Corning microcarriers are offered untreated or with the following surface treatments
- Corning Synthemax™ II substrate creates a synthetic surface on the microcarriers for stem cell expansion
- For use with MSC and iPS/ES applications. (Animal Origin Free)
- Sterility Assurance Level (SAL) of 10−6
- Nonpyrogenic
- Available as closed system solutions
- Corning microcarriers are available sterile and ready to use in 1 to 10 g vials. This format provides flexibility to select the amount of beads to use.
- During the mature phase of process development and into manufacturing, closed system packaging would be necessary to maintain sterility and ease bioreactor loading.
Specifications
Specifications
| For Use With (Application) | MSC and iPS/ES Applications |
| Sterility | Sterile |
| Surface Treatment | Synthemax™ II |
| Product Type | Microcarrier |
| Material | USP Class VI Polystyrene |
| Quantity | 1 g |