missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 10 Mouse anti-Human, Clone: 1B1C3, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | Syntaxin 10 |
|---|---|
| Clone | 1B1C3 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16809055
|
Proteintech
66821-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16899045
|
Proteintech
66821-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SNARE involved in vesicular transport from the late endosomes to the trans-Golgi network.Specifications
| Syntaxin 10 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| hsyn10, STX10, SYN10, syntaxin 10 | |
| STX10 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1B1C3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O60499 | |
| 8677 | |
| Syntaxin 10 Fusion Protein Ag27861 | |
| Primary | |
| -20°C | |
| STX10 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title