Learn More
Agilent BioTek Synergy LC Empty Cube
Shop All Agilent BioTek Products
Click to view available options
:
empty cube
Description
- Empty cube
- Used for custom cubes with EX/EM filters and dichroics
Specifications
Specifications
| For Use With (Equipment) | Synergy LX |
| For Use With (Application) | ELISA, Fluorescence ELISA, Protein Quantification, Nucleic Acid Quantification |