missing translation for 'onlineSavingsMsg'
Learn More

SYNE2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25265-1-AP-20UL

 View more versions of this product

Product Code. 16843845

  • 1710.33 NOK / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Plays a role in pre-mRNA alternative splicing by regulating spliceosome assembly.
Specifications

Specifications

SYNE2
Polyclonal
Unconjugated
SYNE2
KIAA1011, Nesprin 2, NUA, NUANCE, Protein NUANCE, SYNE 2, SYNE2
Rabbit
Antigen Affinity Chromatography
RUO
23224
-20°C
Liquid
Western Blot
0.34 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8WXH0
SYNE2
Peptide
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.