missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25151-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the intermediate filament family which contains an N-terminal head domain, followed by a central coiled-coil region and a short C-terminal tail. The protein is highly expressed in skeletal and cardiac muscle. The protein links the dystrophin associated protein complex (DAPC) to desmin filaments in muscle and may have a structural role in striated muscle. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
SYNC | |
Polyclonal | |
Unconjugated | |
SYNC | |
SYNC, SYNC1, SYNCOILIN, Syncoilin 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
68828, 81493 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9EPM5, Q9H7C4 | |
SYNC | |
SYNC Fusion Protein Ag18565 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |