missing translation for 'onlineSavingsMsg'
Learn More

Synaptotagmin-12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55015-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16886445

  • 3468.15 NOK / 150µL
Estimated Shipment: 13-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody is specific to STY12.

This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. Studies of the orthologous gene in rat have shown that the encoded protein selectively modulates spontaneous synaptic-vesicle exocytosis and may also be involved in regulating calcium independent secretion in nonneuronal cells. Alternative splicing results in multiple transcript variants. The gene has previously been referred to as synaptotagmin XI but has been renamed synaptotagmin XII to be standard with mouse and rat official nomenclature.
Specifications

Specifications

Synaptotagmin-12
Polyclonal
Unconjugated
SYT12
SRG1, Synaptotagmin 12, synaptotagmin XII, Synaptotagmin-12, SYT11, SYT12, SytXII
Rabbit
Antigen Affinity Chromatography
RUO
171180, 191595, 91683
-20°C
Liquid
Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P97610, Q8IV01, Q920N7
Syt12
Peptide
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers