Learn More
Synaptophysin Mouse, anti-Human, Unconjugated, Clone: 4E12C4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67864-1-IG-20UL
Additional Details : Weight : 0.00020kg
 View more versions of this product
Product Code. 17231536
  • 1611.84 NOK / 20µL
Estimated Shipment: 22-05-2024
Description
Synaptophysin (SYP, also known as major synaptic vesicle protein p38) is a 38-kDa integral membrane glycoprotein that regulates synaptic vesicle endocytosis. It is the most abundant synaptic vesicle protein by mass. Synaptophysin is present in neuroendocrine cells and neurons that participate in synaptic transmission. Synaptophysin is a useful marker for identification of neuroendocrine cells and neoplasms. (PMID: 3010302; 21658579)
Specifications
Specifications
Synaptophysin
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
4E12C4
Unconjugated
20 μL
6855