Learn More
Synaptophysin Mouse, anti-Human, Unconjugated, Clone: 4E12C4, Proteintech
Shop All Proteintech ProductsDescription
Synaptophysin (SYP, also known as major synaptic vesicle protein p38) is a 38-kDa integral membrane glycoprotein that regulates synaptic vesicle endocytosis. It is the most abundant synaptic vesicle protein by mass. Synaptophysin is present in neuroendocrine cells and neurons that participate in synaptic transmission. Synaptophysin is a useful marker for identification of neuroendocrine cells and neoplasms. (PMID: 3010302; 21658579)
Specifications
Specifications
| Antigen | Synaptophysin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Clone | 4E12C4 |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 150 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 6855 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.