Learn More
KARTELL Sylicon Conical Stopper
Product Code. p-200004202
Click to view available options
Diameter (Metric) Top:
9 mm
13 mm
18 mm
23 mm
26 mm
42 mm
45 mm
48 mm
54 mm
60 mm
Unit Size:
Pack of 10
Pack of 5
Product Code. 10514631
Brand: KARTELL 0380000
Only null left
Add to basket
Add to basket
The Sylicon Conical Stopper creates a seal in bottles and flasks.
  • Autoclavable.
  • SUITABLE FOR FOODSTUFF.
  • MATERIAL: SYLICON
Specifications
Autoclavable Autoclavable
Color White
Diameter (Metric) Bottom 6 mm
Diameter (Metric) Outer 9 mm
Diameter (Metric) Top 9 mm
Material Silicone