Learn More
KARTELL Sylicon Conical Stopper
Click to view available options
Diameter (Metric) Top:
9 mm
13 mm
18 mm
23 mm
26 mm
42 mm
45 mm
48 mm
54 mm
60 mm
Unit Size:
Pack of 10
Pack of 5
Description
- Autoclavable.
- SUITABLE FOR FOODSTUFF.
- MATERIAL: SYLICON
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | White |
| Diameter (Metric) Bottom | 48 mm |
| Diameter (Metric) Outer | 60 mm |
| Diameter (Metric) Top | 60 mm |
| Material | Silicone |
| Sterility | Non-sterile |