Description
- Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
- Swivel and fixed position clamps hold apparatus near the lab-frame or stand, and have a built-in integral holder for attaching to lab-frames or apparatus
- Available as single or dual-adjust in nickel-plated zinc or stainless steel
- Non-slip vinyl sleeves, and fiberglass covers for temperatures above 100°C (included with all 2 and 3-prong clamps)
Specifications
Specifications
| Type | Multi-purpose Clamp |
| Material | Nickel-plated |
| Includes | Non-slip vinyl sleeves and fiberglass covers |
| Description | Clamp, Multi Purpose, CLM-SWIVL3DZM |
| Length (English) | 7.01 in. |
| Length (Metric) | 178 mm |
| Size | Medium |
| Size (Metric) Grip | 0 to 69 mm |
| Jaw Type | 3-Prong Dual Adjust |