Learn More
Hettich™ Swing-out Rotor for ROTOFIX 32 A and Universal 320/320 R Centrifuge, 8-place
Designed for an extended range of applications
Brand: Hettich™ 1611
Additional Details : Weight : 0.01000kg
Description
- Swing-out Rotor, 8-place
- For use with Rotofix and Universal centrifuges
- Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
- ROTOFIX Centrifuges are ideally equipped for tasks in clinical chemistry, it can also play a vital role in cytology
Specifications
Swing-out Rotor | |
4,000 rpm | |
107 mm | |
45° |
2,415 x g | |
135 mm | |
ROTOFIX 32 A and Universal 320/320 R Centrifuge | |
8 |