Learn More
Hettich™ Swing-out Rotor for ROTIXA 500 RS Centrifuge, 4-place
Description
- Swing-out rotor, 4-place
- For use with ROTIXA 500 RS Centrifuge
Specifications
Specifications
| Type | Swing-out Rotor |
| Max. RCF | 5,252 x g |
| Max. Speed | 4,500 rpm |
| Max. Tube Length | 115 mm |
| Max. Tube Diameter | 40 mm |
| For Use With (Equipment) | ROTIXA 500 RS Centrifuge |
| No. of Places | 4 |