Learn More
Hettich Lab Technology™ Swing-out Rotor for ROTIXA 50 S Centrifuge, 2-place
Product Code. 10445922
Change view
Click to view available options
Capacity:
20 Plates/Racks per Rotor
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
10445922 20 Plates/Racks per Rotor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10445922 Supplier Hettich Lab Technology™ Supplier No. 4282
 Restricted Item
Estimated Shipment: 11-12-2026
Designed for use with ROTIXA 50 S Centrifuge. These rotors are ideal for high sample throughput in medical, research and quality control laboratories.
  • Swing-out Rotor, 2-place
  • For Deep Well Plates
  • For ROTIXA 50 S Centrifuge
  • ROTIXA models are the centrifuges of choice for medical routine laboratories, as well as research and quality control laboratories in the life science industry.
  • ROTIXA centrifuges feature the extended S control panel.
  • Ideal for high sample throughput.
Specifications
Type Swing-out Rotor
Capacity 20 Plates/Racks per Rotor
Max. RCF 2,652 x g
Max. Speed 3,600 rpm
Radius (Metric) 183 mm
Radius (Metric) Min. 178 mm
For Use With (Equipment) ROTIXA 50 S Centrifuge
Angle 90°
No. of Places 2