Learn More
Hettich Lab Technology™ Swing-out Rotor without Carriers
Product Code. 10787141
Change view
Click to view available options
Max. Speed:
15,000 rpm
4,000 rpm
Max. RCF:
2,039 x g
2,218 x g
2,665 x g
24,400 x g
4,193 x g
Unit Size:
Each
Product Code. Max. Speed Max. RCF unitSize
10787141 4,000 rpm 2,665 x g Each
10700095 4,000 rpm 4,193 x g Each
10648452 4,000 rpm 2,039 x g Each
10094092 4,000 rpm 2,218 x g Each
10422405 15,000 rpm 24,400 x g Each
5 options
Product Code. 10787141
Brand: Hettich Lab Technology™ 1624
Swing-out Rotor without Carriers is suitable for performing rate-zonal and isopycnic centrifugations
  • Available in a variety of place number configurations
  • Configurable with a variety of carriers and carrier buckets
Specifications
Type Swing-out Rotor
Capacity Variety of Carriers for Centrifugable Tubes Including Cyto Chambers
Max. RCF 2,665 x g
Max. Speed 4,000 rpm
For Use With (Equipment) ROTOFIX 32 A and Universal 320/320 R Centrifuges
Angle 90°
No. of Places 4