Learn More
Hettich Lab Technology™ Swing-out Rotor without Carriers
Product Code. 10700095
Click to view available options
Max. Speed:
15,000 rpm
4,000 rpm
Max. RCF:
2,039 x g
2,218 x g
2,665 x g
24,400 x g
4,193 x g
Unit Size:
Each
Product Code. 10700095
Brand: Hettich Lab Technology™ 1494
Swing-out Rotor without Carriers is suitable for performing rate-zonal and isopycnic centrifugations
  • Available in a variety of place number configurations
  • Configurable with a variety of carriers and carrier buckets
Specifications
Type Swing-out Rotor
Capacity Variety of Carriers for Centrifugable Tubes
Max. RCF 4,193 x g
Max. Speed 4,000 rpm
For Use With (Equipment) Universal 320/320 R
Angle 90°
No. of Places 4