Learn More
Hettich Lab Technology™ Swing-out Rotor without Carriers
Description
- Available in a variety of place number configurations
- Configurable with a variety of carriers and carrier buckets
Specifications
Specifications
| Type | Swing-out Rotor |
| Capacity | Variety of Carriers for Centrifugable Tubes Including Cyto Chambers |
| Max. RCF | 2,039 x g |
| Max. Speed | 4,000 rpm |
| For Use With (Equipment) | ROTOFIX 32 A and Universal 320/320 R Centrifuges |
| No. of Places | 6 |