Learn More
Hettich Lab Technology™ Swing-out Rotor without Carriers
Click to view available options
Max. Speed:
15,000 rpm
4,000 rpm
Max. RCF:
2,039 x g
2,218 x g
2,665 x g
24,400 x g
4,193 x g
Unit Size:
Each
Description
- Available in a variety of place number configurations
- Configurable with a variety of carriers and carrier buckets
Specifications
Specifications
| Type | Swing-out Rotor |
| Capacity | Variety of Carriers for Centrifugable Tubes |
| Max. RCF | 24,400 x g |
| Max. Speed | 15,000 rpm |
| For Use With (Equipment) | ROTINA 380/380 R |
| Angle | 90° |
| No. of Places | 4 |