Learn More
Thermo Scientific™ Spring Loaded Transfer Line Nut for ISQ and TSQ
Product Code. p-7163707 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
3-Port SilFlow MCD Device
Swap-IT
Description:
Blanking Ferrule
Chromatography Accessories
Unit Size:
Each
Pack of 5
Product Code. 15297488
Brand: Thermo Scientific™ 290ST414
Only null left
Add to basket
Add to basket
Make column changeovers quick and easy. Use this GC-MS interface system to switch capillary columns without the need to vent your mass spectrometer (MS).
TRUSTED_SUSTAINABILITY
Specifications
Description Blanking Ferrule
For Use With (Equipment) 3-Port SilFlow MCD Device