Learn More
MedLine Cotton Swabs in Polypropylene Tubes
Product Code. 11512483
Change view
Click to view available options
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
11512483 Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11512483 Supplier MedLine Supplier No. 300261.2
  • ONLINE EXCLUSIVE PRICE
    1655.00 NOK
    1218.63 NOK / Pack of 500
    Save 436.37 NOK
    26% Off
Estimated Shipment: 09-12-2026
Use these swabs for taking samples. Swabs in Polypropylene Tubes have a round, compact and shred resistant head.
  • Sterile dry swabs supplied in shockproof round bottom polypropylene tube
  • Sterilized by ethylene oxide
  • Produced under strict ISO 9002/EN 29002 standards
  • CE marked No.98 06 0119, Class II A
Specifications
Includes All Swabs Have a Round, Compact and Shred Resistant Head for Sample Taking.
Certifications/Compliance ISO 9002/EN 29002,CE Marked No.98 06 0119
Color Plain
Diameter (Metric) Tip 5 mm, 2.5mm
Grade Class II A
Head Type Swabs Have a Round, Compact and Shred-Resistant Head for Sample Taking
Length (Metric) 150 mm, 150mm
Quantity 500/Pk, 500Pack
Sterility Sterile
Handle Material Polystyrene Shaft
Product Type Tube Swab
Tip Material Cotton
No. per Pack 500