Learn More
SVL™ Sealing Rings for Sliding Joints
Product Code. p-8002836
Change view
Click to view available options
Diameter (Metric) Inside:
11.6 to 12.4 mm
13.6 to 14.4 mm
15.6 to 16.4 mm
17.6 to 18.4 mm
19.5 to 20.5 mm
21.5 to 22.5 mm
29.2 to 30.8 mm
31.2 to 32.8 mm
4.0 to 4.4 mm
5.6 to 6.4 mm
6.6 to 7.4 mm
7.6 to 8.4 mm
9.6 to 10.4 mm
Unit Size:
Pack of 10
Product Code. Diameter (Metric) Inside unitSize
10105674 6.6 to 7.4 mm Pack of 10
12517816 17.6 to 18.4 mm Pack of 10
10287774 31.2 to 32.8 mm Pack of 10
12607245 13.6 to 14.4 mm Pack of 10
10739365 21.5 to 22.5 mm Pack of 10
10442205 7.6 to 8.4 mm Pack of 10
10635905 4.0 to 4.4 mm Pack of 10
10645905 5.6 to 6.4 mm Pack of 10
10146444 15.6 to 16.4 mm Pack of 10
10319944 9.6 to 10.4 mm Pack of 10
10379984 11.6 to 12.4 mm Pack of 10
10014464 19.5 to 20.5 mm Pack of 10
11943889 29.2 to 30.8 mm Pack of 10
13 options
Product Code. 10105674
Brand: SVL 70122
Only null left
Add to basket
Add to basket
Manufactured from silicone rubber and sheathed with PTFE for protection against chemical erosion. SVL™ Sealing Rings for Sliding Joints are ideal for inserting thermometers and glass tubing into an SVL™ system. A wide range of sizes is available for use with tubing from 4.0 to 32.8mm.
  • Sealing rings designed to provide a sliding joint
  • Ideal for inserting thermometers and glass tubing etc. into an SVL™ system
  • Sealing rings manufactured from silicone rubber and sheathed with PTFE for protection against chemical erosion
  • Wide range of sizes for use with tubing from 4.0 to 32.8mm
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inside 6.6 to 7.4 mm
For Use With (Application) Torion joints and sliding joints
For Use With (Equipment) Glass Body 721-02
Joint Size 15
Material Silicone Rubber, PTFE (Sheath)
Type Seal Ring