Learn More
SVL™ Sealing Rings for Sliding Joints
Product Code. 10287774
Click to view available options
Diameter (Metric) Inside:
11.6 to 12.4 mm
13.6 to 14.4 mm
15.6 to 16.4 mm
17.6 to 18.4 mm
19.5 to 20.5 mm
21.5 to 22.5 mm
29.2 to 30.8 mm
31.2 to 32.8 mm
4.0 to 4.4 mm
5.6 to 6.4 mm
6.6 to 7.4 mm
7.6 to 8.4 mm
9.6 to 10.4 mm
Unit Size:
Pack of 10
Product Code. 10287774
Brand: SVL 70129
Only null left
Add to basket
Add to basket
Manufactured from silicone rubber and sheathed with PTFE for protection against chemical erosion. SVL™ Sealing Rings for Sliding Joints are ideal for inserting thermometers and glass tubing into an SVL™ system. A wide range of sizes is available for use with tubing from 4.0 to 32.8mm.
  • Sealing rings designed to provide a sliding joint
  • Ideal for inserting thermometers and glass tubing etc. into an SVL™ system
  • Sealing rings manufactured from silicone rubber and sheathed with PTFE for protection against chemical erosion
  • Wide range of sizes for use with tubing from 4.0 to 32.8mm
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inside 31.2 to 32.8 mm
For Use With (Application) Torion joints and sliding joints
Joint Size 42
Material Silicone Rubber, PTFE (Sheath)
Type Seal Ring