Learn More
SVL™ Sealing Ring for Butt Joint
Product Code. 12681591
Click to view available options
Joint Size:
15
22
30
42
Unit Size:
Each
Pack of 10
Product Code. 12681591
Brand: SVL 70101
Only null left
Add to basket
Add to basket
Manufactured from silicone rubber
  • Silicone rubber and sheathed with PTFE for protection against chemical attack
  • Internal diameter of ring corresponds to the bore of the SVL glass screwthread so that flow is not restricted
TRUSTED_SUSTAINABILITY
Specifications
Description Joint size: 15
For Use With (Application) Butt joints
Joint Size 15
Material Silicone Rubber, PTFE (Sheath)
Type Seal Ring