Learn More
SVL™ PTFE Key
Product Code. 15747967
Change view
Click to view available options
Material:
PTFE
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15747967 PTFE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15747967 Supplier SVL Supplier No. 72053
  • ONLINE EXCLUSIVE PRICE
    675.00 NOK
    581.39 NOK / Pack of 1
    Save 93.61 NOK
    13% Off
Estimated Shipment: 02-03-2027
SVL™ PTFE Key is designed for use with Torion stopcocks.
  • For use with right angle pattern Torion stopcock
  • Screwed into the threaded Torion connector, to open and close the stopcock
Specifications
Type Stopcock Key
Material PTFE
For Use With (Equipment) Torion Stopcock 722-43