Learn More
Rotulex™ Metal Joint Clamps
Product Code. 12059032
Change view
Click to view available options
Size:
Approx. bore size: 15 mm
Approx. bore size: 20 mm
Approx. bore size: 25 mm
Approx. bore size: 5 mm
Approx. bore: 40 mm
Approx. bore: 9 mm
Unit Size:
Each
Pack of 5
Product Code. Size unitSize
12059032 Approx. bore size: 25 mm Each
13191364 Approx. bore size: 20 mm Each
12099111 Approx. bore size: 15 mm Each
12434751 Approx. bore size: 5 mm Each
15557950 Approx. bore: 40 mm Pack of 5
10686475 Approx. bore: 9 mm Each
6 options
Product Code. 12059032
Brand: SVL RC41
Only null left
Add to basket
Add to basket
Spring-loaded metal clips. Rotulex™ Metal Joint Clamps are designed to secure corresponding Rotulex™ glassware systems with ball and socket style joints.
  • Suitable for high temperature work
  • System includes male ball joint, PTFE covered O-ring and female cup joint which are secured with the joint clamp
  • Components are ordered separately
  • Available in a variety of sizes
TRUSTED_SUSTAINABILITY
Specifications
Type Joint Clamp
Material Metal
For Use With (Equipment) Rotulex™ spherical joint system
Size Approx. bore size: 25 mm