Learn More
SVL™ Glass Screwthreads
Product Code. 15527750
Change view
Click to view available options
Diameter (Metric) Outer:
22.5 mm
32 mm
44.1 mm
Unit Size:
Pack of 10
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
15527750 44.1 mm Pack of 50
10136014 22.5 mm Pack of 10
15507750 22.5 mm Pack of 50
15517750 32 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15527750 Supplier SVL™ Supplier No. 70304
  • ONLINE EXCLUSIVE PRICE
    18030.00 NOK
    15930.84 NOK / Pack of 50
    Save 2099.16 NOK
    11% Off
Estimated Shipment: 23-03-2027
For use by glassblowers to construct apparatus with SVR™ joints
  • For use by Glassblowers to construct apparatus with SVL™ joints
  • Borosilicate glass for highest resistance to thermal shock and chemical corrosion
Specifications
Material Borosilicate Glass (3.3)
Diameter (English) Outer 1.7 in.
Diameter (Metric) Outer 44.1 mm
Overall Length (English) 6.49 in.
Overall Length (Metric) 165 mm
Thickness (Metric) 3.2 mm
For Use With (Application) For use by Glassblowers to construct apparatus with SVL™ joints
Product Type Junction Tube