Learn More
SVL™ Glass Screwthreads
Product Code. 15507750
Change view
Click to view available options
Diameter (Metric) Outer:
22.5 mm
32 mm
44.1 mm
Unit Size:
Pack of 10
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
15507750 22.5 mm Pack of 50
10136014 22.5 mm Pack of 10
15517750 32 mm Pack of 50
15527750 44.1 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15507750 Supplier SVL Supplier No. 70302
  • ONLINE EXCLUSIVE PRICE
    3465.00 NOK
    3054.91 NOK / Pack of 50
    Save 410.09 NOK
    11% Off
Estimated Shipment: 22-02-2027
For use by glassblowers to construct apparatus with SVR™ joints
  • For use by Glassblowers to construct apparatus with SVL™ joints
  • Borosilicate glass for highest resistance to thermal shock and chemical corrosion
Specifications
Material Borosilicate Glass
Diameter (Metric) Outer 22.5 mm
Overall Length (English) 6.49 in.
Overall Length (Metric) 165 mm
Thickness (Metric) 1.8 mm
For Use With (Application) For use by Glassblowers to construct apparatus with SVL™ joints
Product Type Junction Tube