Learn More
SVL™ Glass Screwthreads
Product Code. 10136014
Change view
Click to view available options
Diameter (Metric) Outer:
22.5 mm
32 mm
44.1 mm
Unit Size:
Pack of 10
Pack of 50
Product Code. Diameter (Metric) Outer unitSize
10136014 22.5 mm Pack of 10
15507750 22.5 mm Pack of 50
15517750 32 mm Pack of 50
15527750 44.1 mm Pack of 50
4 options
Product Code. 10136014
Brand: SVL 70302
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use by glassblowers to construct apparatus with SVR™ joints
  • For use by Glassblowers to construct apparatus with SVL™ joints
  • Borosilicate glass for highest resistance to thermal shock and chemical corrosion
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass
Diameter (Metric) Outer 22.5 mm
Overall Length (English) 6.49 in.
Overall Length (Metric) 165 mm
Thickness (Metric) 1.8 mm
For Use With (Application) For use by Glassblowers to construct apparatus with SVL™ joints
Product Type Junction Tube