Learn More
Quickfit™ Claisen Still Head Adapter
Product Code. 12048493
missing translation for 'orderingAttributeHoverText'
Joint Type:
Condenser: 19/26, Flask: 24/29
Condenser: 24/29, Flask: 24/29
Size:
Socket: 14/23
missing translation for 'unitSize'
Each
Pack of 5
Product Code. 12048493
missing translation for 'mfr': Quickfit SH6/33
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
PTFE joint sleeves eliminate the possibility of joint seizure
  • Similar to the SH4: still heads but with extra socket to accept dropping funnel or stirrer
TRUSTED_SUSTAINABILITY
Specifications
Type Claisen Still Head
Material Borosilicate Glass
Standard Taper Joint 24/49 (Condenser), 24/29 (Flask)
Joint Type Condenser: 24/29, Flask: 24/29
Size Socket: 14/23