Learn More
SVCT2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27019-1-AP-150UL
This item is not returnable.
View return policy
Description
The SVCT2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human SVCT2. This antibody recognizes human, mouse, rat antigen. The SVCT2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| SVCT2 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| 150 μL | |
| 9962 | |
| IgG |