Learn More
SVCT2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27019-1-AP-150UL
 View more versions of this product
Product Code. 17511356
  • 4836.10 NOK / 150µL
Description
The SVCT2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human SVCT2. This antibody recognizes human, mouse, rat antigen. The SVCT2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
SVCT2
Polyclonal
Rabbit
Primary
Human, Mouse, Rat
Western Blot, Immunohistochemistry
Unconjugated
150 μL
9962