Learn More
uvex suXXeed Safety Glasses
Product Code. 17727679 Shop All Uvex Products
Change view
Click to view available options
Lens Tint:
Clear
Gray (23%)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint Unit Size
17727679 Clear Each
17737679 Gray (23%) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17727679 Supplier uvex Supplier No. 9181265
  • ONLINE EXCLUSIVE PRICE
    152.00 NOK
    146.00 NOK / Pack of 1
    Save 6.00 NOK
    3% Off
View all promotions
Estimated Shipment: 18-12-2026
The Uvex suXXeed protects you all day at work and in many situations.
Sporty safety glasses with a dynamic design that appeals.
General features
  • Sporty safety glasses with a dynamic design that appeals
  • Robust polycarbonate lens with proven uvex supravision coating technology – permanently anti-fog and scratch-resistant lens
  • Optional extra frame provides greater protection against particles, dust and dirt
  • Metal-free
Protection features
  • UV400 – 100% protection against dangerous UVA and UVB radiation to 400 nm
Comfort features
  • Ergonomically shaped arms with soft ear pieces ensure a comfortable and secure fit without creating pressure points
  • Soft, flexible nose pad adapts to the wearer and ensures a comfortable, pressure-free fit
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Chemical-resistant
Lens Coating Anti-fog, Scratch-resistant
Color Anthracite, Blue
Certifications/Compliance (Europe) EN 166, EN 170
Lens Tint Clear
Product Type Ergonomic Safety Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard