Learn More
Poulten Selfe™ Suspended-Level Viscometers
Product Code. 13161043
Change view
Click to view available options
Size:
1
2
2A
3
3A
4
4A
5
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
13161043 1 Each
13181043 2 Each
13191043 2A Each
13121053 4 Each
13111053 3A Each
13131053 4A Each
13101053 3 Each
13141053 5 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 13161043 Supplier Poulten Selfe™ Supplier No. 1625/01
  • ONLINE EXCLUSIVE PRICE
    2535.00 NOK
    2202.59 NOK / Pack of 1
    Save 332.41 NOK
    13% Off
View all promotions
 Restricted Item
Estimated Shipment: 04-01-2027
Suitable for transparent liquids. With UKAS certificate of calibration.
  • Type BS/IP/SL, IP71.Suitable for transparent liquids. Nominal overall length 330mm. Approximate sample size 22mL ­to 40mL. Available without certificate of calibration, please enquire.
  • With UKAS certificate of calibration.
Specifications
Product Type Suspended-Level Viscometer
Viscosity Range 3.5 to 10 cSt
Certifications/Compliance UKAS
Size 1
Volume (Metric) 22 to 40 mL